The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006187.3 | NP_006178.2 | 13 - 14 | 2967 | 97 |
XM_005253889.3 | XP_005253946.1 | 13 - 14 | 2888 | 97 | |
XM_017019363.1 | XP_016874852.1 | 13 - 14 | 2735 | 97 | |
GenBank mRNA | AB044545.1 | - | 13 - 14 | 3048 | 97 |
AF063613.1 | - | 13 - 14 | 2903 | 97 | |
AK292949.1 | - | 13 - 14 | 2937 | 97 | |
BC113746.1 | - | 13 - 14 | 2942 | 97 |