The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006187.3 | NP_006178.2 | 6 - 7 | 1469 | 68 |
XM_005253889.3 | XP_005253946.1 | 6 - 7 | 1390 | 68 | |
XM_017019363.1 | XP_016874852.1 | 6 - 7 | 1390 | 68 | |
GenBank mRNA | AB044545.1 | - | 6 - 7 | 1550 | 68 |
AF063613.1 | - | 6 - 7 | 1405 | 68 | |
AK292949.1 | - | 6 - 7 | 1439 | 68 | |
BC113746.1 | - | 6 - 7 | 1444 | 68 | |
BM474470.1 | - | 1 - 2 | 167 | 68 |