Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OAS3
Entrez Gene ID
4940
Gene Name
2'-5'-oligoadenylate synthetase 3
Gene Aliases
p100, p100OAS
Chromosome Location
Chr.12: 112938433 - 112973251 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006187.3 NP_006178.2 9 - 10 2178 84
XM_005253889.3 XP_005253946.1 9 - 10 2099 84
XM_017019363.1 XP_016874852.1 9 - 10 2099 84
GenBank mRNA AB044545.1 - 9 - 10 2259 84
AF063613.1 - 9 - 10 2114 84
AK292949.1 - 9 - 10 2148 84
BC113746.1 - 9 - 10 2153 84

View Full Product Details