Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INCENP
Entrez Gene ID
3619
Gene Name
inner centromere protein
Gene Aliases
-
Chromosome Location
Chr.11: 62123973 - 62155474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040694.1 NP_001035784.1 18 - 19 2747 100
NM_020238.2 NP_064623.2 17 - 18 2735 100
XM_006718533.2 XP_006718596.1 18 - 19 2721 100
XM_011544995.2 XP_011543297.1 18 - 19 2754 100
XM_011544996.2 XP_011543298.1 18 - 19 2742 100
XM_011544997.2 XP_011543299.1 17 - 18 2702 100
XM_011544998.2 XP_011543300.1 17 - 18 2730 100
GenBank mRNA AY714053.1 - 17 - 18 2545 100
BC098576.1 - 18 - 19 2747 100

View Full Product Details