The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005257.5 | NP_005248.2 | 5 - 6 | 1801 | 86 |
GenBank mRNA | AK223547.1 | - | 4 - 5 | 1730 | 86 |
D87811.1 | - | 4 - 5 | 1342 | 86 | |
U66075.1 | - | 4 - 5 | 1427 | 86 | |
X95701.1 | - | 5 - 6 | 1795 | 86 |