Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
Chromosome Location
Chr.10: 99659506 - 99711245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020354.3 NP_065087.1 13 - 13 5006 89
GenBank mRNA AK123516.1 - 1 - 1 612 89
AL713774.1 - 1 - 1 906 89
BX648264.1 - 1 - 1 2077 89

View Full Product Details