The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289067.1 | NP_001275996.1 | 10 - 11 | 1188 | 129 |
NM_001289068.1 | NP_001275997.1 | 10 - 11 | 1143 | 129 | |
NM_001289071.1 | NP_001276000.1 | 11 - 12 | 1232 | 129 | |
NM_001289073.1 | NP_001276002.1 | 9 - 10 | 1123 | 129 | |
NM_018063.4 | NP_060533.2 | 10 - 11 | 1188 | 129 | |
GenBank mRNA | AB102716.1 | - | 11 - 12 | 1077 | 129 |
AB102717.1 | - | 9 - 10 | 968 | 129 | |
AB102723.1 | - | 10 - 11 | 1033 | 129 | |
AB113248.1 | - | 10 - 11 | 1033 | 129 | |
AB113249.1 | - | 10 - 11 | 1033 | 129 | |
AF155827.1 | - | 10 - 11 | 1138 | 129 | |
AK314485.1 | - | 10 - 11 | 1177 | 129 | |
BX538033.1 | - | 10 - 11 | 1127 | 129 |