Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HELLS
Entrez Gene ID
3070
Gene Name
helicase, lymphoid-specific
Gene Aliases
ICF4, LSH, Nbla10143, PASG, SMARCA6
Chromosome Location
Chr.10: 94545767 - 94613905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289067.1 NP_001275996.1 11 - 12 1391 127
NM_001289068.1 NP_001275997.1 11 - 12 1346 127
NM_001289069.1 NP_001275998.1 11 - 12 1295 127
NM_001289071.1 NP_001276000.1 12 - 13 1435 127
NM_001289073.1 NP_001276002.1 10 - 11 1326 127
NM_001289074.1 NP_001276003.1 12 - 13 1310 127
NM_018063.4 NP_060533.2 11 - 12 1391 127
GenBank mRNA AB102716.1 - 12 - 13 1280 127
AB102717.1 - 10 - 11 1171 127
AB102718.1 - 11 - 12 1140 127
AB102722.1 - 12 - 13 1155 127
AB102723.1 - 11 - 12 1236 127
AB113248.1 - 11 - 12 1236 127
AB113249.1 - 11 - 12 1236 127
AF155827.1 - 11 - 12 1341 127
AK314485.1 - 11 - 12 1380 127
BX538033.1 - 11 - 12 1330 127

View Full Product Details