Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HELLS
Entrez Gene ID
3070
Gene Name
helicase, lymphoid-specific
Gene Aliases
ICF4, LSH, Nbla10143, PASG, SMARCA6
Chromosome Location
Chr.10: 94545767 - 94613905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289067.1 NP_001275996.1 20 - 21 2544 163
NM_001289068.1 NP_001275997.1 19 - 20 2361 163
NM_001289069.1 NP_001275998.1 19 - 20 2310 163
NM_001289070.1 NP_001275999.1 17 - 18 2112 163
NM_001289071.1 NP_001276000.1 20 - 21 2450 163
NM_001289072.1 NP_001276001.1 17 - 18 2016 163
NM_001289073.1 NP_001276002.1 18 - 19 2341 163
NM_001289074.1 NP_001276003.1 20 - 21 2325 163
NM_001289075.1 NP_001276004.1 18 - 19 2209 163
NM_018063.4 NP_060533.2 19 - 20 2406 163
GenBank mRNA AB074174.1 - 7 - 8 854 163
AB102716.1 - 20 - 21 2295 163
AB102717.1 - 18 - 19 2186 163
AB102718.1 - 19 - 20 2155 163
AB102719.1 - 17 - 18 1861 163
AB102720.1 - 18 - 19 2054 163
AB102721.1 - 17 - 18 1957 163
AB102722.1 - 20 - 21 2170 163
AB102723.1 - 20 - 21 2389 163
AB113248.1 - 19 - 20 2164 163
AF155827.1 - 19 - 20 2356 163
AK001201.1 - 5 - 6 564 163
AK314485.1 - 19 - 20 2395 163
AY007108.1 - 7 - 8 863 163
BC015477.1 - 6 - 7 689 163
BC029381.1 - 7 - 8 794 163
BC030963.1 - 8 - 9 1009 163
BC031004.1 - 8 - 9 1009 163
BX538033.1 - 19 - 20 2345 163

View Full Product Details