Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HELLS
Entrez Gene ID
3070
Gene Name
helicase, lymphoid-specific
Gene Aliases
ICF4, LSH, Nbla10143, PASG, SMARCA6
Chromosome Location
Chr.10: 94545767 - 94613905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289067.1 NP_001275996.1 8 - 9 863 98
NM_001289068.1 NP_001275997.1 8 - 9 818 98
NM_001289069.1 NP_001275998.1 8 - 9 863 98
NM_001289070.1 NP_001275999.1 8 - 9 863 98
NM_001289071.1 NP_001276000.1 9 - 10 907 98
NM_001289072.1 NP_001276001.1 8 - 9 863 98
NM_001289073.1 NP_001276002.1 7 - 8 798 98
NM_001289074.1 NP_001276003.1 8 - 9 863 98
NM_001289075.1 NP_001276004.1 8 - 9 863 98
NM_018063.4 NP_060533.2 8 - 9 863 98
GenBank mRNA AB102716.1 - 9 - 10 752 98
AB102717.1 - 7 - 8 643 98
AB102718.1 - 8 - 9 708 98
AB102719.1 - 8 - 9 708 98
AB102720.1 - 8 - 9 708 98
AB102721.1 - 8 - 9 708 98
AB102722.1 - 8 - 9 708 98
AB102723.1 - 8 - 9 708 98
AB113248.1 - 8 - 9 708 98
AB113249.1 - 8 - 9 708 98
AF155827.1 - 8 - 9 813 98
AK308331.1 - 8 - 9 783 98
AK314485.1 - 8 - 9 852 98
BX538033.1 - 8 - 9 802 98

View Full Product Details