Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
Chromosome Location
Chr.10: 63167221 - 63522075 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282948.1 NP_001269877.1 6 - 7 1242 130
NM_001318153.1 NP_001305082.1 6 - 7 1246 130
NM_001318154.1 NP_001305083.1 7 - 8 1184 130
NM_001322252.1 NP_001309181.1 6 - 7 1239 130
NM_001322254.1 NP_001309183.1 5 - 6 1136 130
NM_001322258.1 NP_001309187.1 6 - 7 1078 130
NM_032776.2 NP_116165.1 7 - 8 1353 130
NR_134512.1 4 - 5 1022 130
XM_011539503.1 XP_011537805.1 6 - 7 846 130
XM_011539508.2 XP_011537810.1 6 - 7 846 130
XM_017015897.1 XP_016871386.1 6 - 7 841 130
XM_017015898.1 XP_016871387.1 7 - 8 1034 130
XM_017015899.1 XP_016871388.1 7 - 8 934 130
XM_017015900.1 XP_016871389.1 7 - 8 1338 130
XM_017015901.1 XP_016871390.1 6 - 7 881 130
XM_017015902.1 XP_016871391.1 6 - 7 1142 130
XM_017015903.1 XP_016871392.1 5 - 6 690 130
GenBank mRNA AK027280.1 - 7 - 8 1198 130
BC143722.1 - 6 - 7 1085 130
BX537954.1 - 4 - 5 987 130
EF068222.1 - 9 - 10 1310 130

View Full Product Details