The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321080.1 | NP_001308009.1 | 9 - 10 | 1067 | 116 |
NM_001321083.1 | NP_001308012.1 | 10 - 11 | 1014 | 116 | |
NM_001321084.1 | NP_001308013.1 | 9 - 10 | 931 | 116 | |
NM_020771.3 | NP_065822.2 | 10 - 11 | 1199 | 116 | |
NR_104424.1 | 11 - 12 | 1293 | 116 | ||
XM_017011113.1 | XP_016866602.1 | 10 - 11 | 1066 | 116 | |
XM_017011114.1 | XP_016866603.1 | 9 - 10 | 885 | 116 | |
XM_017011115.1 | XP_016866604.1 | 9 - 10 | 1041 | 116 | |
XM_017011116.1 | XP_016866605.1 | 9 - 10 | 1096 | 116 | |
XM_017011118.1 | XP_016866607.1 | 7 - 8 | 619 | 116 | |
XM_017011119.1 | XP_016866608.1 | 9 - 10 | 902 | 116 | |
XM_017011120.1 | XP_016866609.1 | 8 - 9 | 1017 | 116 | |
XM_017011121.1 | XP_016866610.1 | 8 - 9 | 984 | 116 | |
XM_017011122.1 | XP_016866611.1 | 9 - 10 | 736 | 116 | |
GenBank mRNA | AK291760.1 | - | 10 - 11 | 1124 | 116 |
BC034982.2 | - | 10 - 11 | 1163 | 116 |