The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170794.1 | NP_001164265.1 | 2 - 3 | 547 | 81 |
NM_021813.3 | NP_068585.1 | 4 - 5 | 718 | 81 | |
XM_011536039.2 | XP_011534341.1 | 3 - 4 | 629 | 81 | |
XM_011536040.2 | XP_011534342.1 | 2 - 3 | 578 | 81 | |
XM_017011165.1 | XP_016866654.1 | 3 - 4 | 644 | 81 | |
XM_017011166.1 | XP_016866655.1 | 4 - 5 | 722 | 81 | |
GenBank mRNA | AB208889.1 | - | 2 - 3 | 184 | 81 |
AF357835.1 | - | 1 - 2 | 43 | 81 | |
AJ271878.1 | - | 4 - 5 | 552 | 81 | |
DB077323.1 | - | 2 - 3 | 482 | 81 |