The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016021.2 | NP_057105.2 | 2 - 3 | 430 | 85 |
XM_011535887.1 | XP_011534189.1 | 2 - 3 | 417 | 85 | |
XM_011535888.2 | XP_011534190.1 | 2 - 3 | 447 | 85 | |
GenBank mRNA | AF161502.1 | - | 2 - 3 | 125 | 85 |
AJ245898.1 | - | 2 - 3 | 161 | 85 | |
AJ420578.1 | - | 1 - 2 | 79 | 85 | |
AK223464.1 | - | 3 - 4 | 169 | 85 | |
AK290574.1 | - | 2 - 3 | 272 | 85 | |
AK300629.1 | - | 3 - 4 | 175 | 85 | |
BC013973.1 | - | 2 - 3 | 392 | 85 | |
BF058850.1 | - | 3 - 4 | 349 | 85 |