Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTLL8
Entrez Gene ID
164714
Gene Name
tubulin tyrosine ligase like 8
Gene Aliases
-
Chromosome Location
Chr.22: 50018575 - 50058227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017029176.1 XP_016884665.1 13 - 14 2363 143

View Full Product Details