The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005576.3 | NP_005567.2 | 4 - 5 | 1831 | 75 |
XM_017022179.1 | XP_016877668.1 | 4 - 5 | 847 | 75 | |
GenBank mRNA | AK303982.1 | - | 5 - 6 | 1435 | 75 |
AK314222.1 | - | 4 - 5 | 1829 | 75 | |
BC015090.2 | - | 4 - 5 | 1810 | 75 | |
BC068542.1 | - | 4 - 5 | 1831 | 75 | |
L21186.1 | - | 4 - 5 | 1810 | 75 |