The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278469.1 | NP_001265398.1 | 28 - 29 | 4586 | 102 |
NM_001278472.1 | NP_001265401.1 | 29 - 30 | 4664 | 102 | |
NM_015026.2 | NP_055841.2 | 28 - 29 | 4586 | 102 | |
XM_017019041.1 | XP_016874530.1 | 29 - 30 | 4589 | 102 | |
XM_017019043.1 | XP_016874532.1 | 30 - 31 | 4663 | 102 | |
XM_017019044.1 | XP_016874533.1 | 30 - 31 | 4665 | 102 | |
XM_017019045.1 | XP_016874534.1 | 31 - 32 | 4333 | 102 | |
XM_017019046.1 | XP_016874535.1 | 25 - 26 | 3758 | 102 | |
GenBank mRNA | AB017814.1 | - | 28 - 29 | 4311 | 102 |
AB028963.3 | - | 29 - 30 | 4557 | 102 | |
AL833066.2 | - | 28 - 29 | 4581 | 102 | |
AL834320.1 | - | 22 - 23 | 3544 | 102 | |
BC151241.1 | - | 29 - 30 | 4580 | 102 | |
BX537415.1 | - | 28 - 29 | 4584 | 102 |