Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
-
Chromosome Location
Chr.4: 87160103 - 87220608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292003.1 NP_001278932.1 5 - 6 1484 140
NM_001292006.1 NP_001278935.1 5 - 6 1213 140
NM_001292007.1 NP_001278936.1 4 - 5 892 140
NM_020803.4 NP_065854.3 5 - 6 1441 140
NR_120447.1 3 - 4 705 140
GenBank mRNA AB037799.2 - 4 - 5 1124 140
AK128827.1 - 5 - 6 1484 140
BC041384.2 - 5 - 6 1391 140
BC041901.1 - 3 - 4 561 140
BX640727.1 - 5 - 6 1351 140
BX640744.1 - 5 - 6 1142 140
BX647454.1 - 5 - 6 1376 140
BX648367.1 - 3 - 4 409 140
CR749265.1 - 4 - 5 810 140

View Full Product Details