The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003830.3 | NP_003821.1 | 4 - 5 | 881 | 114 |
XM_011527438.2 | XP_011525740.1 | 4 - 5 | 835 | 114 | |
XM_017027419.1 | XP_016882908.1 | 4 - 5 | 835 | 114 | |
GenBank mRNA | AF170484.1 | - | 4 - 5 | 778 | 114 |
AK225753.1 | - | 4 - 5 | 802 | 114 | |
BC029896.1 | - | 4 - 5 | 783 | 114 | |
U71383.1 | - | 4 - 5 | 884 | 114 |