Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SELENBP1
Entrez Gene ID
8991
Gene Name
selenium binding protein 1
Gene Aliases
HEL-S-134P, LPSB, SBP56, SP56, hSBP
Chromosome Location
Chr.1: 151364302 - 151372734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258288.1 NP_001245217.1 7 - 8 829 102
NM_001258289.1 NP_001245218.1 8 - 9 1224 102
NM_003944.3 NP_003935.2 8 - 9 1015 102
GenBank mRNA AK023875.1 - 5 - 6 1825 102
AK296661.1 - 8 - 9 1193 102
AK298355.1 - 8 - 9 880 102
AK303815.1 - 7 - 8 795 102
AK315643.1 - 8 - 9 985 102
BC009084.1 - 8 - 9 969 102
BC032997.1 - 6 - 7 1400 102
CR456852.1 - 7 - 8 922 102
EU794660.1 - 8 - 9 969 102

View Full Product Details