The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142548.1 | NP_001136020.1 | 2 - 3 | 119 | 122 |
NM_003579.3 | NP_003570.2 | 1 - 2 | 716 | 122 | |
GenBank mRNA | AK292611.1 | - | 2 - 3 | 99 | 122 |
BC121059.1 | - | 1 - 2 | 224 | 122 | |
BC121060.1 | - | 1 - 2 | 224 | 122 | |
BM464345.1 | - | - | 691 | 123 | |
X97795.1 | - | 1 - 2 | 105 | 122 |