Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VRK1
Entrez Gene ID
7443
Gene Name
vaccinia related kinase 1
Gene Aliases
PCH1, PCH1A
Chromosome Location
Chr.14: 96797309 - 96881614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003384.2 NP_003375.1 5 - 6 484 98
XM_006720247.3 XP_006720310.1 5 - 6 522 98
XM_011537132.1 XP_011535434.1 4 - 5 383 98
XM_017021622.1 XP_016877111.1 4 - 5 383 98
XM_017021624.1 XP_016877113.1 5 - 6 522 98
XM_017021625.1 XP_016877114.1 5 - 6 563 98
XM_017021626.1 XP_016877115.1 5 - 6 522 98
GenBank mRNA AB000449.1 - 5 - 6 453 98
AK290110.1 - 5 - 6 472 98
BC103761.1 - 5 - 6 462 98
BC112075.1 - 5 - 6 452 98
BC113510.1 - 5 - 6 452 98

View Full Product Details