Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VRK1
Entrez Gene ID
7443
Gene Name
vaccinia related kinase 1
Gene Aliases
PCH1, PCH1A
Chromosome Location
Chr.14: 96797309 - 96881614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003384.2 NP_003375.1 6 - 7 590 72
XM_006720247.3 XP_006720310.1 6 - 7 628 72
XM_011537132.1 XP_011535434.1 5 - 6 489 72
XM_017021622.1 XP_016877111.1 5 - 6 489 72
XM_017021624.1 XP_016877113.1 6 - 7 628 72
XM_017021625.1 XP_016877114.1 6 - 7 669 72
XM_017021626.1 XP_016877115.1 6 - 7 628 72
GenBank mRNA AB000449.1 - 6 - 7 559 72
AK290110.1 - 6 - 7 578 72
BC103761.1 - 6 - 7 568 72
BC112075.1 - 6 - 7 558 72
BC113510.1 - 6 - 7 558 72

View Full Product Details