The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015156.3 | NP_055971.2 | 4 - 5 | 727 | 91 |
GenBank mRNA | AF155595.1 | - | 4 - 5 | 490 | 91 |
BC051003.1 | - | 4 - 5 | 725 | 91 | |
BC064495.1 | - | 4 - 5 | 725 | 91 | |
D31888.1 | - | 4 - 5 | 232 | 91 |