The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114.4 | NP_001105.1 | 15 - 16 | 2201 | 87 |
NM_001286057.1 | NP_001272986.1 | 16 - 17 | 2249 | 87 | |
XM_011522835.2 | XP_011521137.1 | 16 - 17 | 2302 | 87 | |
XM_011522836.2 | XP_011521138.1 | 16 - 17 | 2318 | 87 | |
XM_011522837.2 | XP_011521139.1 | 16 - 17 | 2247 | 87 | |
XM_011522838.2 | XP_011521140.1 | 16 - 17 | 2451 | 87 | |
XM_011522839.2 | XP_011521141.1 | 16 - 17 | 2278 | 87 | |
XM_011522840.2 | XP_011521142.1 | 16 - 17 | 2285 | 87 | |
XM_011522842.2 | XP_011521144.1 | 16 - 17 | 2348 | 87 | |
XM_017022895.1 | XP_016878384.1 | 16 - 17 | 2318 | 87 | |
XM_017022896.1 | XP_016878385.1 | 13 - 14 | 1464 | 87 | |
XM_017022897.1 | XP_016878386.1 | 13 - 14 | 1483 | 87 | |
XM_017022898.1 | XP_016878387.1 | 16 - 17 | 2318 | 87 | |
XM_017022899.1 | XP_016878388.1 | 16 - 17 | 2318 | 87 | |
GenBank mRNA | AK093706.1 | - | 17 - 18 | 2199 | 87 |
BC039891.1 | - | 13 - 14 | 1837 | 87 | |
BC126271.1 | - | 15 - 16 | 2013 | 87 | |
D25538.1 | - | 15 - 16 | 2198 | 87 |