The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020884.4 | NP_065935.3 | 41 - 42 | 5984 | 71 |
XM_006723840.3 | XP_006723903.1 | 39 - 40 | 5625 | 71 | |
XM_011528941.2 | XP_011527243.1 | 39 - 40 | 5649 | 71 | |
XM_011528947.2 | XP_011527249.1 | 33 - 34 | 6022 | 71 | |
XM_011528948.2 | XP_011527250.1 | 33 - 34 | 5181 | 71 | |
XM_017027986.1 | XP_016883475.1 | 40 - 41 | 5685 | 71 | |
XM_017027987.1 | XP_016883476.1 | 35 - 36 | 5200 | 71 | |
GenBank mRNA | AB040945.2 | - | 39 - 40 | 5693 | 71 |
BC007808.1 | - | 14 - 15 | 2671 | 71 | |
BC151242.1 | - | 39 - 40 | 5705 | 71 |