The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032865.5 | NP_116254.4 | 12 - 13 | 2170 | 60 |
XM_005257744.1 | XP_005257801.1 | 12 - 13 | 2246 | 60 | |
XM_017025236.1 | XP_016880725.1 | 11 - 12 | 2171 | 60 | |
GenBank mRNA | AF370421.1 | - | 12 - 13 | 2166 | 60 |
AF417488.1 | - | 12 - 13 | 2132 | 60 | |
AK027856.1 | - | 10 - 11 | 1520 | 60 | |
BC013373.2 | - | 3 - 4 | 282 | 60 | |
BC018706.1 | - | 11 - 12 | 1616 | 60 |