The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010986.2 | NP_001010986.1 | 16 - 17 | 1779 | 78 |
NM_173694.4 | NP_775965.2 | 16 - 17 | 1779 | 78 | |
XM_017029439.1 | XP_016884928.1 | 17 - 18 | 1785 | 78 | |
XM_017029440.1 | XP_016884929.1 | 17 - 18 | 2456 | 78 | |
XM_017029441.1 | XP_016884930.1 | 18 - 19 | 1865 | 78 | |
XM_017029442.1 | XP_016884931.1 | 17 - 18 | 2006 | 78 | |
XM_017029443.1 | XP_016884932.1 | 17 - 18 | 1784 | 78 | |
XM_017029444.1 | XP_016884933.1 | 16 - 17 | 2396 | 78 | |
XM_017029445.1 | XP_016884934.1 | 17 - 18 | 1786 | 78 | |
XM_017029446.1 | XP_016884935.1 | 16 - 17 | 1716 | 78 | |
XM_017029447.1 | XP_016884936.1 | 17 - 18 | 1989 | 78 | |
XM_017029448.1 | XP_016884937.1 | 17 - 18 | 2121 | 78 | |
XM_017029449.1 | XP_016884938.1 | 16 - 17 | 1727 | 78 | |
GenBank mRNA | AJ580093.1 | - | 16 - 17 | 1781 | 78 |
AJ580094.1 | - | 16 - 17 | 1781 | 78 | |
AK125986.1 | - | 6 - 7 | 769 | 78 | |
AK131262.1 | - | 1 - 2 | 55 | 78 | |
BC132728.1 | - | 16 - 17 | 1717 | 78 | |
BC132730.1 | - | 16 - 17 | 1717 | 78 | |
BX648320.1 | - | 3 - 4 | 221 | 78 |