Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGCZ
Entrez Gene ID
137868
Gene Name
sarcoglycan zeta
Gene Aliases
ZSG1
Chromosome Location
Chr.8: 14084835 - 15238283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322880.1 NP_001309809.1 2 - 3 889 110
NM_139167.3 NP_631906.2 2 - 3 889 110
GenBank mRNA AY028700.1 - 1 - 2 190 110
AY502063.1 - 2 - 3 229 110
BC069459.1 - 1 - 2 190 110

View Full Product Details