The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001942.3 | NP_001933.2 | 2 - 3 | 292 | 93 |
GenBank mRNA | AF097935.1 | - | 2 - 3 | 293 | 93 |
BX476267.1 | - | 2 - 3 | 265 | 93 | |
X56654.1 | - | 2 - 3 | 157 | 93 |