The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001752.3 | NP_001743.1 | 2 - 3 | 330 | 122 |
GenBank mRNA | AK291585.1 | - | 2 - 3 | 330 | 122 |
AK301577.1 | - | 1 - 2 | 100 | 122 | |
AK315350.1 | - | 2 - 3 | 314 | 122 | |
AY028632.1 | - | 2 - 3 | 241 | 122 | |
BC110398.1 | - | 2 - 3 | 319 | 122 | |
BC112217.1 | - | 2 - 3 | 279 | 122 | |
BC112219.1 | - | 2 - 3 | 279 | 122 | |
X04076.1 | - | 2 - 3 | 311 | 122 |