The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001752.3 | NP_001743.1 | 3 - 4 | 437 | 60 |
GenBank mRNA | AK291585.1 | - | 3 - 4 | 437 | 60 |
AK301577.1 | - | 2 - 3 | 207 | 60 | |
AK310184.1 | - | 2 - 3 | 262 | 60 | |
AK315350.1 | - | 3 - 4 | 421 | 60 | |
AY028632.1 | - | 3 - 4 | 348 | 60 | |
BC110398.1 | - | 3 - 4 | 426 | 60 | |
BC112217.1 | - | 3 - 4 | 386 | 60 | |
BC112219.1 | - | 3 - 4 | 386 | 60 | |
X04076.1 | - | 3 - 4 | 418 | 60 |