The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001752.3 | NP_001743.1 | 5 - 6 | 672 | 76 |
GenBank mRNA | AK291585.1 | - | 5 - 6 | 672 | 76 |
AK301577.1 | - | 4 - 5 | 442 | 76 | |
AK310184.1 | - | 4 - 5 | 497 | 76 | |
AK315350.1 | - | 5 - 6 | 656 | 76 | |
AY028632.1 | - | 5 - 6 | 583 | 76 | |
BC110398.1 | - | 5 - 6 | 661 | 76 | |
BC112217.1 | - | 5 - 6 | 621 | 76 | |
BC112219.1 | - | 5 - 6 | 621 | 76 | |
X04076.1 | - | 5 - 6 | 653 | 76 |