The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001752.3 | NP_001743.1 | 6 - 7 | 807 | 123 |
GenBank mRNA | AK291585.1 | - | 6 - 7 | 807 | 123 |
AK301577.1 | - | 5 - 6 | 577 | 123 | |
AK310184.1 | - | 5 - 6 | 632 | 123 | |
AK315350.1 | - | 6 - 7 | 791 | 123 | |
BC110398.1 | - | 6 - 7 | 796 | 123 | |
BC112217.1 | - | 6 - 7 | 756 | 123 | |
BC112219.1 | - | 6 - 7 | 756 | 123 | |
X04076.1 | - | 6 - 7 | 788 | 123 |