The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001752.3 | NP_001743.1 | 7 - 8 | 984 | 133 |
GenBank mRNA | AK291585.1 | - | 7 - 8 | 984 | 133 |
AK301577.1 | - | 6 - 7 | 754 | 133 | |
AK310184.1 | - | 6 - 7 | 809 | 133 | |
AK315350.1 | - | 7 - 8 | 968 | 133 | |
BC110398.1 | - | 7 - 8 | 973 | 133 | |
BC112217.1 | - | 7 - 8 | 933 | 133 | |
BC112219.1 | - | 7 - 8 | 933 | 133 | |
X04076.1 | - | 7 - 8 | 965 | 133 |