The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001752.3 | NP_001743.1 | 8 - 9 | 1146 | 61 |
GenBank mRNA | AK291585.1 | - | 8 - 9 | 1146 | 61 |
AK301577.1 | - | 7 - 8 | 916 | 61 | |
AK315350.1 | - | 8 - 9 | 1130 | 61 | |
AY028632.1 | - | 8 - 9 | 1057 | 61 | |
BC027300.1 | - | 1 - 2 | 160 | 61 | |
BC110398.1 | - | 8 - 9 | 1135 | 61 | |
BC112217.1 | - | 8 - 9 | 1095 | 61 | |
BC112219.1 | - | 8 - 9 | 1095 | 61 | |
X04076.1 | - | 8 - 9 | 1127 | 61 |