The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079872.1 | NP_001073341.1 | 3 - 4 | 1095 | 96 |
NM_003588.3 | NP_003579.3 | 5 - 6 | 1262 | 96 | |
XM_005262481.1 | XP_005262538.1 | 5 - 6 | 1262 | 96 | |
XM_006724784.1 | XP_006724847.1 | 4 - 5 | 930 | 96 | |
XM_006724785.2 | XP_006724848.1 | 4 - 5 | 841 | 96 | |
XM_011531399.2 | XP_011529701.1 | 3 - 4 | 337 | 96 | |
XM_011531400.2 | XP_011529702.1 | 3 - 4 | 360 | 96 | |
XM_011531401.1 | XP_011529703.1 | 3 - 4 | 198 | 96 | |
GenBank mRNA | AB014595.2 | - | 3 - 4 | 429 | 96 |
AK299081.1 | - | 3 - 4 | 366 | 96 | |
AK315037.1 | - | 5 - 6 | 1198 | 96 | |
AY365125.1 | - | 3 - 4 | 771 | 96 | |
BC036216.1 | - | 5 - 6 | 1227 | 96 | |
BX537641.1 | - | 3 - 4 | 1217 | 96 | |
BX537787.1 | - | 3 - 4 | 1086 | 96 | |
BX647096.1 | - | 3 - 4 | 1095 | 96 | |
CR936722.1 | - | 3 - 4 | 1095 | 96 |