Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL2
Entrez Gene ID
79368
Gene Name
Fc receptor like 2
Gene Aliases
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
Chromosome Location
Chr.1: 157745733 - 157777472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159488.1 NP_001152960.1 6 - 7 1212 73
NM_030764.3 NP_110391.2 6 - 7 1212 73
XM_011509974.2 XP_011508276.1 6 - 7 1547 73
XM_011509975.2 XP_011508277.1 6 - 7 1549 73
XM_011509976.2 XP_011508278.1 6 - 7 1548 73
XM_017002316.1 XP_016857805.1 6 - 7 1547 73
XM_017002317.1 XP_016857806.1 6 - 7 1350 73
XM_017002319.1 XP_016857808.1 5 - 6 1262 73
GenBank mRNA AF319438.1 - 1 - 2 697 73
AF319439.1 - 1 - 2 535 73
AF390037.1 - 1 - 2 535 73
AF459633.1 - 6 - 7 1200 73
AK301113.1 - 6 - 7 1199 73
AL833194.1 - 1 - 2 3071 73
AY043465.1 - 6 - 7 1199 73
AY358130.1 - 6 - 7 1202 73
BC129836.1 - 1 - 2 537 73

View Full Product Details