The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030764.3 | NP_110391.2 | 8 - 9 | 1363 | 107 |
NR_125358.1 | 5 - 6 | 511 | 107 | ||
XM_011509974.2 | XP_011508276.1 | 8 - 9 | 1698 | 107 | |
XM_011509975.2 | XP_011508277.1 | 8 - 9 | 1700 | 107 | |
XM_011509976.2 | XP_011508278.1 | 8 - 9 | 1699 | 107 | |
XM_017002316.1 | XP_016857805.1 | 8 - 9 | 1698 | 107 | |
XM_017002317.1 | XP_016857806.1 | 8 - 9 | 1501 | 107 | |
XM_017002318.1 | XP_016857807.1 | 7 - 8 | 1421 | 107 | |
XM_017002319.1 | XP_016857808.1 | 7 - 8 | 1413 | 107 | |
GenBank mRNA | AF319438.1 | - | 3 - 4 | 848 | 107 |
AF390037.1 | - | 3 - 4 | 686 | 107 | |
AF459633.1 | - | 8 - 9 | 1351 | 107 | |
AL833194.1 | - | 3 - 4 | 3222 | 107 | |
AY043465.1 | - | 8 - 9 | 1350 | 107 | |
AY358130.1 | - | 8 - 9 | 1353 | 107 | |
BC069185.1 | - | 5 - 6 | 515 | 107 | |
BC129836.1 | - | 3 - 4 | 688 | 107 |