The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145169.2 | NP_660152.1 | 1 - 2 | 192 | 95 |
GenBank mRNA | AF041429.1 | - | 1 - 2 | 126 | 95 |
BC018969.1 | - | 1 - 2 | 97 | 95 | |
BG706927.1 | - | 1 - 2 | 181 | 95 |