The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145169.2 | NP_660152.1 | 7 - 8 | 567 | 142 |
NR_130112.1 | 8 - 9 | 590 | 142 | ||
GenBank mRNA | AF041429.1 | - | 7 - 8 | 501 | 142 |
BC018969.1 | - | 7 - 8 | 472 | 142 | |
BI597156.1 | - | 8 - 9 | 552 | 142 |