The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130851.1 | NP_001124323.1 | 4 - 5 | 409 | 84 |
NM_005192.3 | NP_005183.2 | 5 - 6 | 529 | 84 | |
XM_011536353.2 | XP_011534655.1 | 5 - 6 | 545 | 84 | |
GenBank mRNA | AF213033.1 | - | 4 - 5 | 415 | 84 |
AF213036.1 | - | 4 - 5 | 415 | 84 | |
AF213038.1 | - | 3 - 4 | 295 | 84 | |
AF213039.1 | - | 4 - 5 | 415 | 84 | |
AF213041.1 | - | 4 - 5 | 415 | 84 | |
AF213042.1 | - | 3 - 4 | 295 | 84 | |
AF213044.1 | - | 2 - 3 | 277 | 84 | |
AF213045.1 | - | 4 - 5 | 397 | 84 | |
AF213046.1 | - | 4 - 5 | 415 | 84 | |
AF213047.1 | - | 3 - 4 | 295 | 84 | |
AF213048.1 | - | 2 - 3 | 277 | 84 | |
AF213049.1 | - | 3 - 4 | 295 | 84 | |
AF213051.1 | - | 3 - 4 | 295 | 84 | |
AF213052.1 | - | 4 - 5 | 415 | 84 | |
AK098653.1 | - | 5 - 6 | 529 | 84 | |
AY257474.1 | - | 4 - 5 | 415 | 84 | |
BC064965.1 | - | 4 - 5 | 369 | 84 | |
BT019451.1 | - | 4 - 5 | 415 | 84 | |
CR407666.1 | - | 4 - 5 | 415 | 84 | |
EF560750.1 | - | 5 - 6 | 455 | 84 | |
KP966095.1 | - | 4 - 5 | 446 | 84 | |
L25876.1 | - | 5 - 6 | 476 | 84 | |
L27711.1 | - | 5 - 6 | 466 | 84 | |
U02681.1 | - | 5 - 6 | 446 | 84 |