Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSD3
Entrez Gene ID
23362
Gene Name
pleckstrin and Sec7 domain containing 3
Gene Aliases
EFA6D, EFA6R, HCA67
Chromosome Location
Chr.8: 18527303 - 19086909 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015310.3 NP_056125.3 10 - 11 2319 108
NM_206909.2 NP_996792.1 7 - 8 723 108
XM_017013258.1 XP_016868747.1 10 - 11 3628 108
XM_017013259.1 XP_016868748.1 10 - 11 3606 108
XM_017013260.1 XP_016868749.1 10 - 11 2379 108
XM_017013261.1 XP_016868750.1 9 - 10 2593 108
XM_017013265.1 XP_016868754.1 7 - 8 1129 108
GenBank mRNA AB023159.2 - 7 - 8 704 108
AF243495.2 - 8 - 9 729 108
AF519767.1 - 8 - 9 677 108
AK091317.1 - 10 - 11 2319 108
AK315099.1 - 7 - 8 751 108
AL832778.1 - 9 - 10 2186 108
BC075044.2 - 7 - 8 701 108
BC075045.2 - 8 - 9 704 108
GU727648.1 - 10 - 11 2219 108

View Full Product Details