The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308045.1 | NP_001294974.1 | 4 - 5 | 2002 | 74 |
NM_152699.4 | NP_689912.2 | 4 - 5 | 2002 | 74 | |
XM_005269310.2 | XP_005269367.1 | 4 - 5 | 2002 | 74 | |
XM_005269312.2 | XP_005269369.1 | 4 - 5 | 2002 | 74 | |
XM_011512544.2 | XP_011510846.1 | 5 - 6 | 2047 | 74 | |
XM_011512545.2 | XP_011510847.1 | 4 - 5 | 2002 | 74 | |
XM_017005870.1 | XP_016861359.1 | 5 - 6 | 2002 | 74 | |
GenBank mRNA | AF335474.1 | - | 3 - 4 | 859 | 74 |
AK124389.1 | - | 3 - 4 | 975 | 74 | |
AK302305.1 | - | 4 - 5 | 2002 | 74 | |
AK313764.1 | - | 4 - 5 | 1975 | 74 | |
BC000958.2 | - | 3 - 4 | 1099 | 74 | |
BC008589.1 | - | 3 - 4 | 1099 | 74 | |
BC030705.2 | - | 4 - 5 | 1951 | 74 |