The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142296.1 | NP_001135768.1 | 2 - 3 | 266 | 102 |
XM_005266314.3 | XP_005266371.1 | 2 - 3 | 257 | 102 | |
XM_005266315.3 | XP_005266372.1 | 2 - 3 | 111 | 102 |