Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL32
Entrez Gene ID
64983
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
HSPC283, L32mt, MRP-L32, bMRP-59b
Chromosome Location
Chr.7: 42932205 - 42937857 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK090551.1 - 2 - 3 2426 77

View Full Product Details