Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF24
Entrez Gene ID
11237
Gene Name
ring finger protein 24
Gene Aliases
G1L
Chromosome Location
Chr.20: 3927309 - 4015591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134337.2 NP_001127809.1 2 - 3 302 135
NM_001134338.2 NP_001127810.1 3 - 4 371 135
NM_001321749.1 NP_001308678.1 2 - 3 538 135
NM_007219.4 NP_009150.1 2 - 3 594 135
XM_011529145.2 XP_011527447.1 4 - 5 617 135
XM_011529146.2 XP_011527448.1 3 - 4 547 135
XM_011529147.2 XP_011527449.1 4 - 5 625 135
XM_017027620.1 XP_016883109.1 3 - 4 639 135
XM_017027621.1 XP_016883110.1 3 - 4 408 135
XM_017027622.1 XP_016883111.1 3 - 4 483 135
GenBank mRNA AL096778.1 - 2 - 3 392 135
BC000213.2 - 3 - 4 325 135
BC039584.1 - 2 - 3 221 135
BT007406.1 - 1 - 2 140 135
CV811405.1 - 2 - 3 280 135

View Full Product Details