Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF24
Entrez Gene ID
11237
Gene Name
ring finger protein 24
Gene Aliases
G1L
Chromosome Location
Chr.20: 3927309 - 4015591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134337.2 NP_001127809.1 4 - 5 396 125
NM_001134338.2 NP_001127810.1 5 - 6 465 125
NM_001321749.1 NP_001308678.1 4 - 5 632 125
NM_007219.4 NP_009150.1 4 - 5 688 125
XM_011529145.2 XP_011527447.1 6 - 7 711 125
XM_011529146.2 XP_011527448.1 5 - 6 641 125
XM_011529147.2 XP_011527449.1 6 - 7 719 125
XM_017027620.1 XP_016883109.1 5 - 6 733 125
XM_017027621.1 XP_016883110.1 5 - 6 502 125
XM_017027622.1 XP_016883111.1 5 - 6 577 125
GenBank mRNA AL079313.1 - 2 - 3 104 125
AL096778.1 - 4 - 5 486 125
BC000213.2 - 5 - 6 419 125
BC039584.1 - 4 - 5 315 125
BT007406.1 - 3 - 4 234 125
CV811405.1 - 4 - 5 374 125

View Full Product Details