The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006282.3 | NP_006273.1 | 11 - 11 | 126 | |
XM_005260530.2 | XP_005260587.1 | 11 - 11 | 126 | ||
XM_005260531.3 | XP_005260588.1 | 12 - 12 | 126 | ||
XM_005260533.2 | XP_005260590.1 | 12 - 12 | 126 | ||
GenBank mRNA | AB209315.1 | - | 13 - 13 | 126 | |
AK027088.1 | - | 2 - 2 | 126 | ||
BC042861.1 | - | 1 - 1 | 126 | ||
BU633556.1 | - | 1 - 1 | 89 | 126 | |
BU689980.1 | - | 1 - 1 | 79 | 126 |