Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INTS6
Entrez Gene ID
26512
Gene Name
integrator complex subunit 6
Gene Aliases
DBI-1, DDX26, DDX26A, DICE1, HDB, INT6, Notchl2
Chromosome Location
Chr.13: 51353171 - 51453236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039937.1 NP_001035026.1 7 - 8 865 117
NM_001306091.1 NP_001293020.1 8 - 9 1641 117
NM_012141.2 NP_036273.1 6 - 7 1357 117
XM_011535040.2 XP_011533342.1 6 - 7 1454 117
XM_011535041.1 XP_011533343.1 5 - 6 1270 117
XM_017020540.1 XP_016876029.1 6 - 7 910 117
GenBank mRNA AF097645.1 - 6 - 7 1219 117
AK074946.1 - 4 - 5 447 117
AK096696.1 - 3 - 4 577 117
AK128795.1 - 4 - 5 512 117
AK296801.1 - 6 - 7 1254 117
AK303869.1 - 5 - 6 664 117
AK304218.1 - 6 - 7 1254 117
AL833524.1 - 7 - 8 933 117
BC013358.2 - 6 - 7 1235 117
BC039829.1 - 6 - 7 1231 117

View Full Product Details